2-Fluoro-5-(trifluoromethyl)benzyl alcohol 80%

Supplier: Ambeed
Warning

Synonyms: (2-Fluoro-5-(trifluoromethyl)phenyl)methanol

A210170-10G A210170-25G
77237-378EA 58 USD
77237-378 77237-380
2-Fluoro-5-(trifluoromethyl)benzyl alcohol 80%
2-Fluoro-5-(trifluoromethyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₆F₄O
MDL Number: MFCD00061167
CAS Number: 207974-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR