N-[3-[3-(Dimethylamino)acryloyl]phenyl]-N-ethylacetamide 98%

Supplier: Ambeed
Warning

Synonyms: N-(3-(3-(Dimethylamino)acryloyl)phenyl)-N-ethylacetamide

A210053-100G A210053-1G A210053-25G A210053-5G
77275-130EA 207 USD
77275-130 77275-132 77275-134 77275-136
N-[3-[3-(Dimethylamino)acryloyl]phenyl]-N-ethylacetamide 98%
N-[3-[3-(Dimethylamino)acryloyl]phenyl]-N-ethylacetamide

Precaución: for Research Use Only

Formula: C₁₅H₂₀N₂O₂
MDL Number: MFCD04117958
CAS Number: 96605-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR