2,2'-Diiodobiphenyl 95%

Supplier: Ambeed
Danger

Synonyms: 2,2'-Diiodo-1,1'-biphenyl

A209890-100MG A209890-1G
77249-050EA 47.38 USD
77249-050 77249-052
2,2'-Diiodobiphenyl 95%
2,2'-Diiodobiphenyl

Precaución: for Research Use Only

Formula: C₁₂H₈I₂
MW: 406.0009 g/mol
MDL Number: MFCD00017622
CAS Number: 2236-52-4
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR