1-Acetyl-4-fluoronaphthalene 98%

Supplier: Ambeed
Warning

Synonyms: 4'-Fluoro-1'-acetonaphthone, 1-(4-Fluoronaphthalen-1-yl)ethanone, 1-(4-Fluoro-1-naphthyl)ethanone

A209875-10G A209875-1G
77287-932EA 109.18 USD
77287-932 77287-934
1-Acetyl-4-fluoronaphthalene 98%
1-Acetyl-4-fluoronaphthalene

Precaución: for Research Use Only

Formula: C₁₂H₉FO
MW: 188.20 g/mol
MDL Number: MFCD00134475
CAS Number: 316-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR