KPT-335 ≥98%

Supplier: Ambeed
Warning

Synonyms: Verdinexor, (Z)-3-[3-[3,5-bis(trifluoromethyl)phenyl]-1,2,4-triazol-1-yl]-N'-pyridin-2-ylprop-2-enehydrazide, (Z)-3-(3-(3,5-Bis(trifluoromethyl)phenyl)-1H-1,2,4-triazol-1-yl)-N'-(pyridin-2-yl)acrylohydrazide

A209806-1MG A209806-25MG A209806-50MG A209806-5MG
77237-356EA 11.33 USD
77237-356 77237-358 77237-360 77237-362
KPT-335 ≥98%
KPT-335

Precaución: for Research Use Only

Formula: C₁₈H₁₂F₆N₆O
MW: 442.32 g/mol
MDL Number: MFCD28167840
CAS Number: 1392136-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR