2-Amino-4-(methoxycarbonyl)benzoic acid 97%

Supplier: Ambeed
Warning

A209800-100MG A209800-1G A209800-250MG A209800-5G
77249-042EA 19 USD
77249-042 77249-044 77249-046 77249-048
2-Amino-4-(methoxycarbonyl)benzoic acid 97%
2-Amino-4-(methoxycarbonyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₉NO₄
MDL Number: MFCD08690068
CAS Number: 85743-02-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR