2-Bromoindolo[3,2,1-jk]carbazole 97%

Supplier: Ambeed
Warning

A209528-100MG A209528-1G A209528-250MG A209528-5G
77237-336EA 32.96 USD
77237-336 77237-338 77237-340 77615-758
2-Bromoindolo[3,2,1-jk]carbazole 97%
2-Bromoindolo[3,2,1-jk]carbazole

Precaución: for Research Use Only

Formula: C₁₈H₁₀BrN
MW: 320.19 g/mol
MDL Number: MFCD30187281
CAS Number: 1174032-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR