4-(4-Aminophenoxy)pyridine 95%

Supplier: Ambeed
Warning

Synonyms: 4-(Pyridin-4-yloxy)aniline

A209500-100MG A209500-1G A209500-250MG
77287-922EA 45.32 USD
77287-922 77287-924 77287-926
4-(4-Aminophenoxy)pyridine 95%
4-(4-Aminophenoxy)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂O
MW: 186.21 g/mol
MDL Number: MFCD06825431
CAS Number: 102877-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR