Methyl 4-(methylsulfonyl)benzoate 98%

Supplier: Ambeed
Warning

A209211-1G A209211-250MG A209211-25G A209211-5G
77287-910EA 28 USD
77287-910 77287-912 77287-914 77287-916
Methyl 4-(methylsulfonyl)benzoate 98%
Methyl 4-(methylsulfonyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₁₀O₄S
MDL Number: MFCD00100037
CAS Number: 22821-70-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR