5-Bromo-1H-1,2,4-triazole 98%

Supplier: Ambeed
Warning

A209143-100G A209143-10G A209143-1G A209143-25G A209143-500G A209143-5G
77275-110EA 59 USD
77275-110 77275-112 77275-114 77275-116 77275-118 77275-120
5-Bromo-1H-1,2,4-triazole 98%
5-Bromo-1H-1,2,4-triazole

Precaución: for Research Use Only

Formula: C₂H₂BrN₃
MDL Number: MFCD09055371
CAS Number: 7343-33-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR