2-Fluoro-5-methoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

A209110-100MG A209110-10G A209110-250MG A209110-1G A209110-25G
77275-104EA 5 USD
77275-104 77275-106 77275-108 77615-754 77748-310
2-Fluoro-5-methoxyphenylboronic acid 98%
2-Fluoro-5-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BFO₃
MDL Number: MFCD04974115
CAS Number: 406482-19-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR