3-(tert-Butyldimethylsiloxy)glutaric anhydride 98%

Supplier: Ambeed
Warning

Synonyms: 3-(tert-Butyldimethylsilyloxy)glutaric anhydride

A208658-1G A208658-5G
77275-076EA 19.57 USD
77275-076 77748-308
3-(tert-Butyldimethylsiloxy)glutaric anhydride 98%
3-(tert-Butyldimethylsiloxy)glutaric anhydride

Precaución: for Research Use Only

Formula: C₁₁H₂₀O₄Si
MW: 244.362 g/mol
MDL Number: MFCD00075235
CAS Number: 91424-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR