17-Amino-10-oxo-3,6,12,15-tetraoxa-9-azaheptadecan-1-oic acid 98%

Supplier: Ambeed
Warning

Synonyms: AEEA-AEEA

A208527-1G A208527-25G A208527-5G
77249-014EA 59.74 USD
77249-014 77249-016 77249-018
17-Amino-10-oxo-3,6,12,15-tetraoxa-9-azaheptadecan-1-oic acid 98%
17-Amino-10-oxo-3,6,12,15-tetraoxa-9-azaheptadecan-1-oic acid

Precaución: for Research Use Only

Formula: C₁₂H₂₄N₂O₇
MW: 308.33 g/mol
MDL Number: MFCD13184942
CAS Number: 1143516-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR