2,3,4,5-Tetrafluoroaniline 97%
Supplier: Ambeed
|
Danger
|
A208417-1G
A208417-250MG
A208417-5G
A208417-25G
A208417-10G
77287-888EA
14.42
USD
77287-888
77287-890
77287-892
77615-752
77748-307
2,3,4,5-Tetrafluoroaniline 97%
2,3,4,5-Tetrafluoroaniline
Precaución: for Research Use Only
Formula:
C₆H₃F₄N MW: 165.09 g/mol |
MDL Number:
MFCD00025153 CAS Number: 5580-80-3 UN: 2810 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...