4-Ethyl-3-nitrobenzoic acid 98%

Supplier: Ambeed
Warning

A208368-100G A208368-10G A208368-1G A208368-25G A208368-5G
77275-066EA 244.11 USD
77275-066 77275-068 77275-070 77275-072 77275-074
4-Ethyl-3-nitrobenzoic acid 98%
4-Ethyl-3-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉NO₄
MW: 195.17 g/mol
MDL Number: MFCD08460111
CAS Number: 103440-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR