(3,5-Difluorophenyl)acetonitrile 97%

Supplier: Ambeed
Warning

A208329-1G A208329-25G A208329-5G
77237-320EA 18.54 USD
77237-320 77237-322 77237-324 77237-326
(3,5-Difluorophenyl)acetonitrile 97%
(3,5-Difluorophenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₈H₅F₂N
MDL Number: MFCD00061278
CAS Number: 122376-76-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR