Methyl-3-aminopyrazine-2-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-aminopyrazine-3-carboxylate

A208317-100G A208317-25G A208317-5G
77275-050EA 65 USD
77275-050 77275-052 77275-054 77275-056 77275-058
Methyl-3-aminopyrazine-2-carboxylate 95%
Methyl 3-aminopyrazine-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇N₃O₂
MDL Number: MFCD00010097
CAS Number: 16298-03-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR