(R)-5,5'-Bis(diphenylphosphino)-2,2,2',2'-tetrafluoro-4,4'-bi-1,3-benzodioxole ≥98%, ee 99%

Supplier: Ambeed
Warning

A208281-100MG A208281-1G A208281-250MG A208281-5G
77249-006EA 102 USD
77249-006 77249-008 77249-010 77249-012
(R)-5,5'-Bis(diphenylphosphino)-2,2,2',2'-tetrafluoro-4,4'-bi-1,3-benzodioxole ≥98%, ee 99%
(R)-5,5'-Bis(diphenylphosphino)-2,2,2',2'-tetrafluoro-4,4'-bi-1,3-benzodioxole

Precaución: for Research Use Only

Formula: C₃₈H₂₄F₄O₄P₂
MDL Number: MFCD05861607
CAS Number: 503538-69-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR