2-Chloro-3,4-dimethoxybenzoic acid 98%

Supplier: Ambeed
Warning

A208046-1G A208046-25G A208046-5G A208046-100G
77260-986EA 7 USD
77260-986 77260-988 77260-990 77748-306
2-Chloro-3,4-dimethoxybenzoic acid 98%
2-Chloro-3,4-dimethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉ClO₄
MDL Number: MFCD00084944
CAS Number: 52009-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR