Benzyl 4-(((methylsulfonyl)oxy)methyl)piperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: 1-Cbz-4-(methylsulphonyloxymethyl)piperidine

A208009-1G A208009-250MG A208009-5G
77287-868EA 22.66 USD
77287-868 77287-870 77287-872
Benzyl 4-(((methylsulfonyl)oxy)methyl)piperidine-1-carboxylate 95%
Benzyl 4-(((methylsulfonyl)oxy)methyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₅S
MW: 327.396 g/mol
MDL Number: MFCD06409219
CAS Number: 159275-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR