4-Fluorobenzo[d]thiazole 95%

Supplier: Ambeed
Warning

A207983-100MG A207983-1G A207983-250MG A207983-5G
77237-304EA 25.75 USD
77237-304 77237-306 77237-308 77746-278
4-Fluorobenzo[d]thiazole 95%
4-Fluorobenzo[d]thiazole

Precaución: for Research Use Only

Formula: C₇H₄FNS
MW: 153.17 g/mol
MDL Number: MFCD17014583
CAS Number: 1247348-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR