4-(Acetyloxy)-3-methoxybenzoic acid 95%

Supplier: Ambeed
Warning

A207501-100G A207501-10G A207501-25G A207501-5G
77287-828EA 258 USD
77287-828 77287-830 77287-832 77287-834
4-(Acetyloxy)-3-methoxybenzoic acid 95%
4-(Acetyloxy)-3-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₅
MDL Number: MFCD00016515
CAS Number: 10543-12-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR