3-(3-Pyridyl)-L-alanine dihydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: H-3-(3-Pyridyl)-Ala-OH·2HCl, (S)-2-Amino-3-(pyridin-3-yl)propanoic acid dihydrochloride

A207406-10G A207406-1G A207406-250MG
77275-036EA 331 USD
77275-036 77275-038 77275-040
3-(3-Pyridyl)-L-alanine dihydrochloride 95%
3-(3-Pyridyl)-L-alanine dihydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂·2ClH
MDL Number: MFCD00191178
CAS Number: 93960-20-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR