2-((4-Fluorophenyl)sulfonyl)ethanol 97%

Supplier: Ambeed
Danger

A207379-100MG A207379-1G A207379-250MG
77237-288EA 22 USD
77237-288 77237-290 77237-292
2-((4-Fluorophenyl)sulfonyl)ethanol 97%
2-((4-Fluorophenyl)sulfonyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₉FO₃S
MDL Number: MFCD03789137
CAS Number: 28122-13-6
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR