3-(2-(Prop-2-yn-1-yloxy)ethoxy)propanoic acid 97%

Supplier: Ambeed
Warning

Synonyms: Propargyl-PEG2-acid

A207192-100MG A207192-1G A207192-250MG A207192-5G A207192-25G
77275-012EA 13 USD
77275-012 77275-014 77275-016 77275-018 77567-986
3-(2-(Prop-2-yn-1-yloxy)ethoxy)propanoic acid 97%
3-(2-(Prop-2-yn-1-yloxy)ethoxy)propanoic acid

Precaución: For research use only.

Formula: C₈H₁₂O₄
MW: 172.17 g/mol
MDL Number: MFCD29764323
CAS Number: 1859379-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR