6-Hydrazinyl-1H-indazole tetrahydrochloride 95%

Supplier: Ambeed
Warning

A207177-100MG A207177-1G A207177-250MG A207177-5G
77260-962EA 33.99 USD
77260-962 77260-964 77260-966 77260-968
6-Hydrazinyl-1H-indazole tetrahydrochloride 95%
6-Hydrazinyl-1H-indazole tetrahydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₂Cl₄N₄
MW: 294.01 g/mol
MDL Number: MFCD30189418
CAS Number: 1266362-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR