2,3,6,7-Tetrahydrobenzo[1,2-b:4,5-b']difuran 97%

Supplier: Ambeed
Warning

A206955-1G A206955-250MG A206955-5G
77248-982EA 20.6 USD
77248-982 77248-984 77248-986
2,3,6,7-Tetrahydrobenzo[1,2-b:4,5-b']difuran 97%
2,3,6,7-Tetrahydrobenzo[1,2-b:4,5-b']difuran

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₂
MW: 162.18 g/mol
MDL Number: MFCD08436969
CAS Number: 81926-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR