[2,2':6',2''-Terpyridin]-4'-amine 97%

Supplier: Ambeed
Warning

A206891-100MG A206891-250MG
77248-980EA 130 USD
77248-980 77567-984
[2,2':6',2''-Terpyridin]-4'-amine 97%
[2,2':6',2''-Terpyridin]-4'-amine

Precaución: For research use only.

Formula: C₁₅H₁₂N₄
MDL Number: MFCD06796972
CAS Number: 193944-66-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR