4-(Benzyloxy)pyridine 98%

Supplier: Ambeed
Warning

A206838-1G A206838-250MG A206838-5G A206838-10G
77287-820EA 87.55 USD
77287-820 77287-822 77287-824 77748-301
4-(Benzyloxy)pyridine 98%
4-(Benzyloxy)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO
MW: 185.222 g/mol
MDL Number: MFCD08235112
CAS Number: 49826-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR