Heptakis-(6-Mercapto-6-deoxy)-β-Cyclodextrin 99%

Supplier: Ambeed
Warning

A206833-100MG A206833-1G
77260-946EA 32.96 USD
77260-946 77260-948
Heptakis-(6-Mercapto-6-deoxy)-β-Cyclodextrin 99%
Heptakis-(6-Mercapto-6-deoxy)-β-Cyclodextrin

Precaución: for Research Use Only

Formula: C₄₂H₇₀O₂₈S₇
MW: 1247.44 g/mol
MDL Number: MFCD30179373
CAS Number: 160661-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR