2-Chloro-N-(2,6-dichlorophenyl)-N-phenylacetamide 97%

Supplier: Ambeed
Warning

A206630-100MG A206630-10G A206630-1G A206630-250MG A206630-5G
77260-936EA 28 USD
77260-936 77260-938 77260-940 77260-942 77260-944
2-Chloro-N-(2,6-dichlorophenyl)-N-phenylacetamide 97%
2-Chloro-N-(2,6-dichlorophenyl)-N-phenylacetamide

Precaución: for Research Use Only

Formula: C₁₄H₁₀Cl₃NO
MDL Number: MFCD00437681
CAS Number: 15308-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR