2,2-Dimethylvaleric acid 98%

Supplier: Ambeed
Danger

Synonyms: 2,2-Dimethylpentanoic acid

A206455-1G A206455-250MG A206455-25G A206455-5G
77248-964EA 37 USD
77248-964 77248-966 77248-968 77248-970
2,2-Dimethylvaleric acid 98%
2,2-Dimethylvaleric acid

Precaución: for Research Use Only

Formula: C₇H₁₄O₂
MDL Number: MFCD00021686
CAS Number: 1185-39-3
UN: 3265
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR