JNK-IN-8 98%

Supplier: Ambeed
Warning

Synonyms: 3-[[(E)-4-(dimethylamino)but-2-enoyl]amino]-N-[3-methyl-4-[(4-pyridin-3-ylpyrimidin-2-yl)amino]phenyl]benzamide, (E)-3-(4-(dimethylamino)but-2-enamido)-N-(3-methyl-4-(4-(pyridin-3-yl)pyrimidin-2-ylamino)phenyl)benzamide, 3-[4-(dimethylamino)but-2-enamido]-N-(3-methyl-4-{[4-(pyridin-3-yl)pyrimidin-2-yl]amino}phenyl)benzamide, 3-(4-(Dimethylamino)but-2-enamido)-N-(3-methyl-4-((4-(pyridin-3-yl)pyrimidin-2-yl)amino)phenyl)benzamide

A206353-10MG A206353-2MG A206353-50MG A206353-5MG A206353-1MG
77237-266EA 33.99 USD
77237-266 77237-268 77237-270 77237-272 77746-273
JNK-IN-8 98%
JNK-IN-8

Precaución: for Research Use Only

Formula: C₂₉H₂₉N₇O₂
MW: 507.59 g/mol
MDL Number: MFCD22124890
CAS Number: 1410880-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR