SU-5214 97%

Supplier: Ambeed
Warning

Synonyms: 3-(2-Methoxy-benzylidene)-1,3-dihydro-indol-2-one, 3-(2-Methoxybenzylidene)indolin-2-one

A206125-100MG A206125-10MG A206125-1MG A206125-50MG A206125-5MG
77260-918EA 363.59 USD
77260-918 77260-920 77260-922 77260-924 77260-926
SU-5214 97%
SU-5214

Precaución: for Research Use Only

Formula: C₁₆H₁₃NO₂
MW: 251.28 g/mol
MDL Number: MFCD01595259
CAS Number: 186611-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR