Ethyl 4-(1-methyl-1H-pyrazol-4-yl)benzoate 97%

Supplier: Ambeed
Warning

A206019-100MG A206019-250MG
77274-994EA 150 USD
77274-994 77274-996
Ethyl 4-(1-methyl-1H-pyrazol-4-yl)benzoate 97%
Ethyl 4-(1-methyl-1H-pyrazol-4-yl)benzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₄N₂O₂
MDL Number: MFCD31619102
CAS Number: 82525-27-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR