Tris(2-fluorophenyl)phosphine 98%

Supplier: Ambeed
Warning

A205860-100MG A205860-1G A205860-250MG A205860-5G
77248-956EA 15 USD
77248-956 77248-958 77248-960 77248-962
Tris(2-fluorophenyl)phosphine 98%
Tris(2-fluorophenyl)phosphine

Precaución: for Research Use Only

Formula: C₁₈H₁₂F₃P
MDL Number: MFCD00015721
CAS Number: 84350-73-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR