4,5-Dimethyl-1H-pyrazol-3-amine 96%
Supplier: Ambeed
|
Warning
|
A205830-100MG
A205830-10G
A205830-1G
A205830-250MG
A205830-25G
A205830-5G
77287-800EA
10.3
USD
77287-800
77287-802
77287-804
77287-806
77287-808
77287-810
4,5-Dimethyl-1H-pyrazol-3-amine 96%
4,5-Dimethyl-1H-pyrazol-3-amine
Precaución: for Research Use Only
Formula:
C₅H₉N₃ MW: 111.14 g/mol |
MDL Number:
MFCD08272069 CAS Number: 91159-73-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...