N-Methyl-1-(4-(trifluoromethyl)phenyl)ethanamine 97%

Supplier: Ambeed
Danger

A205707-1G A205707-250MG
77248-946EA 330.63 USD
77248-946 77248-948
N-Methyl-1-(4-(trifluoromethyl)phenyl)ethanamine 97%
N-Methyl-1-(4-(trifluoromethyl)phenyl)ethanamine

Precaución: for Research Use Only

Formula: C₁₀H₁₂F₃N
MW: 203.20 g/mol
MDL Number: MFCD09763629
CAS Number: 574731-05-8
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR