Adenine 98%

Supplier: Ambeed
Danger

Synonyms: Purin-6-amine, 7H-Purin-6-amine, 6-Aminopurine

A205648-25G A205648-5G A205648-100G
77274-990EA 15 USD
77274-990 77274-992 77593-340
Adenine 98%
Adenine

Precaución: for Research Use Only

Formula: C₅H₅N₅
MW: 135.13 g/mol
MDL Number: MFCD00041790
CAS Number: 73-24-5
UN: 2811
ADR: 6.1 Ⅲ
Merck Index: 13,00152

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR