Adenine hydrochloride hemihydrate 97%

Supplier: Ambeed
Warning

Synonyms: Purin-6-amine hydrochloride hemihydrate, 1H-Purin-6-amine hydrochloride, 7H-Purin-6-amine hydrochloride hemihydrate, 6-Aminopurine Hydrochloride, Adenine Hydrochloride Hemihydrate, 6-Aminopurine hydrochloride hemihydrate, 7H-Purin-6-amine hydrochloride hydrate (2:2:1)

A205443-100G A205443-100MG A205443-10G A205443-25G A205443-5G
77287-780EA 117 USD
77287-780 77287-782 77287-784 77287-786 77287-788
Adenine hydrochloride hemihydrate 97%
Adenine hydrochloride hemihydrate

Precaución: for Research Use Only

Formula: C₅H₅N₅·HCl·½H₂O
MW: 180.60 g/mol
MDL Number: MFCD11046306
CAS Number: 6055-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR