PNU-282987 98%

Supplier: Ambeed
Warning

Synonyms: (R)-4-Chloro-N-(quinuclidin-3-yl)benzamide hydrochloride, N-[(3R)-1-Azabicyclo[2.2.2]oct-3-yl]-4-chlorobenzamide

A205325-100MG A205325-10MG A205325-1MG A205325-250MG A205325-25MG A205325-50MG
77237-250EA 106.09 USD
77237-250 77237-252 77237-254 77237-256 77237-258 77237-260
PNU-282987 98%
PNU-282987

Precaución: for Research Use Only

Formula: C₁₄H₁₈Cl₂N₂O
MW: 301.211 g/mol
MDL Number: MFCD08703110
CAS Number: 123464-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR