2,4-Dichlorobenzotrifluoride 98%

Supplier: Ambeed
Danger

Synonyms: 2,4-Dichlorobenzotrifluoride, 1,3-Dichloro-4-(trifluoromethyl)benzene, 2,4-Dichloro-1-(trifluoromethyl)benzene

A205087-100G A205087-25G
77248-926EA 10.3 USD
77248-926 77248-928
2,4-Dichlorobenzotrifluoride 98%
2,4-Dichlorobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₃Cl₂F₃
MW: 215 g/mol
MDL Number: MFCD00000582
CAS Number: 320-60-5
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR