Trichloroacetamide 98%

Supplier: Ambeed
Warning

Synonyms: 2,2,2-Trichloroacetamide

A205031-1000G A205031-100G A205031-25G A205031-500G
77248-918EA 169.95 USD
77248-918 77248-920 77248-922 77248-924
Trichloroacetamide 98%
Trichloroacetamide

Precaución: for Research Use Only

Formula: C₂H₂Cl₃NO
MW: 162.4 g/mol
MDL Number: MFCD00008009
CAS Number: 594-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR