2-Chloro-1,6-naphthyridine 95%

Supplier: Ambeed
Warning

A204994-1G A204994-250MG
77260-894EA 562.38 USD
77260-894 77260-896
2-Chloro-1,6-naphthyridine 95%
2-Chloro-1,6-naphthyridine

Precaución: for Research Use Only

Formula: C₈H₅ClN₂
MW: 164.59 g/mol
MDL Number: MFCD11044731
CAS Number: 23616-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR