2-Fluoro-3-iodonitrobenzene 95%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-1-iodo-3-nitrobenzene

A204941-100MG A204941-250MG
77237-246EA 123.6 USD
77237-246 77237-248
2-Fluoro-3-iodonitrobenzene 95%
2-Fluoro-3-iodonitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃FINO₂
MW: 266.996 g/mol
MDL Number: MFCD18396988
CAS Number: 1261782-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR