2-Nitro-p-toluic acid 98%
Supplier: Ambeed
|
Warning
|
A204824-10G
A204824-1G
A204824-250MG
A204824-5G
77287-762EA
22
USD
77287-762
77287-764
77287-766
77287-768
2-Nitro-p-toluic acid 98%
2-Nitro-p-toluic acid
Precaución: for Research Use Only
Formula:
C₈H₇NO₄ |
MDL Number:
MFCD00033899 CAS Number: 27329-27-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...