3,4-Dichlorothiophenol 98%

Supplier: Ambeed
Danger

Synonyms: 1,2-Dichloro-4-mercaptobenzene, 1,2-Dichloro-4-benzenethiol, 3,4-Dichlorobenzenethiol, 3,4-Dichlorobenzene-1-thiol

A204454-1G A204454-25G A204454-5G
77274-960EA 11.33 USD
77274-960 77274-962 77274-964
3,4-Dichlorothiophenol 98%
3,4-Dichlorothiophenol

Precaución: for Research Use Only

Formula: C₆H₄Cl₂S
MW: 179.07 g/mol
MDL Number: MFCD00004840
CAS Number: 5858-17-3
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR