3-Bromo-5-phenylpyridine 98%

Supplier: Ambeed
Warning

A204358-100MG A204358-1G A204358-250MG A204358-5G
77274-952EA 22 USD
77274-952 77274-954 77274-956 77274-958
3-Bromo-5-phenylpyridine 98%
3-Bromo-5-phenylpyridine

Precaución: for Research Use Only

Formula: C₁₁H₈BrN
MDL Number: MFCD04114223
CAS Number: 142137-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR