4-Chlorocinnamic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-(4-Chlorophenyl)acrylic acid

A204281-100G A204281-25G A204281-5G
77260-878EA 45.32 USD
77260-878 77260-880 77260-882
4-Chlorocinnamic acid 98%
4-Chlorocinnamic acid

Precaución: for Research Use Only

Formula: C₉H₇ClO₂
MW: 182.61 g/mol
MDL Number: MFCD00004396
CAS Number: 1615-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR