4-Ethoxy-2,3-difluorophenol 98%

Supplier: Ambeed
Warning

A204149-100G A204149-10G A204149-1G A204149-25G A204149-5G
77237-224EA 278.1 USD
77237-224 77237-226 77237-228 77237-230 77237-232
4-Ethoxy-2,3-difluorophenol 98%
4-Ethoxy-2,3-difluorophenol

Precaución: for Research Use Only

Formula: C₈H₈F₂O₂
MW: 174.14 g/mol
MDL Number: MFCD07368025
CAS Number: 126163-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR